missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPAP2B Monoclonal Antibody (ZV002)
GREENER_CHOICE

Product Code. 10088662
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10088662 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10088662 Supplier Invitrogen™ Supplier No. 391000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Mouse Monoclonal Antibody

PPAP2B Monoclonal Antibody for Western Blot, ICC/IF, ELISA

Human vascular endothelial growth factor produced in E.Coli is a double, non-glycosylated, polypeptide chain containing 165 amino acids and having a molecular mass of 38231 Dalton. The rHuVEGF is purified by proprietary chromatographic techniques.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPAP2B
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone ZV002
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene PLPP3
Gene Accession No. O14495
Gene Alias 1110003O22Rik; 2610002D05Rik; AV025606; D4Bwg0538e; D4Bwg1535e; differentially expressed in rat intestine 42; Dri42; EC 3.1.3.4; ER transmembrane protein Dri 42; lipid phosphate phosphohydrolase 3; Lpp3; MGC15306; PAP2 beta; PAP2B; PAP-2b; PAP2-b; PAP2-beta; phosphatidate phosphohydrolase type 2b; Phosphatidic acid phosphatase 2b; phosphatidic acid phosphatase type 2B; phospholipid phosphatase 3; PLPP3; Ppab2b; PPAP2B; type-2 phosphatidic acid phosphatase-beta; vascular endothelial growth factor and type I collagen inducible; Vascular endothelial growth factor and type I collagen-inducible protein; VCIP
Gene Symbols PLPP3
Host Species Mouse
Immunogen Recombinant protein derived from an internal region of human VCIP.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8613
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.