missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ Transparent Polypropylene Reagent Bottles with NS-Type Stopper

Product Code. 10154032
Change view
Click to view available options
Capacity (Metric):
1000 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10154032 1000 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10154032 Supplier VITLAB™ Supplier No. 100694
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

VITLAB™ Transparent Polypropylene Reagent Bottles with NS-Type Stopper offer good chemical resistance and are ideal for long-term storage of liquids. They are autoclavable at 121°C (2 bar) according to DIN EN 285 (restrictions may apply).

  • Product Type: Reagent Bottle
  • Material: Polypropylene (Bottle, Stopper)
  • Color: Transparent
  • Closure: Stopper (NS 29/32)
  • Includes: Stopper Type A (Square Cap with Red Core)

Specifications

Capacity (Metric) 1000 mL
Color Transparent
Diameter (English) 4.3 in.
Material Polypropylene
Max. Temperature (Metric) Autoclaving: 121°C (2 bar)
No. per Case 1Each
No. per Pack 1Each
Packaging 1Each
Quantity 1Each
Sterility Non-sterile
Closure Color Transparent with Red Core
Certifications/Compliance DIN EN 285 (Restrictions May Apply)
Closure Included Yes
Disposable Reusable
Graduated No
Shape Round
Height (Metric) 213 mm
Product Type Reagent Bottle
Height (English) 8.4 in.
Closure Material Polypropylene
Closure Size NS 29/32
Diameter (Metric) 108 mm
Closure Type Stopper
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.