missing translation for 'onlineSavingsMsg'
Learn More

Marigold™ Powder-free Blue Nitrile Gloves

Product Code. 12445060
Change view
Click to view available options
:
Color: blue; Size: M
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12445060 Color: blue; Size: M
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12445060 Supplier Marigold™ Supplier No. 10198/

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Certifications/Compliance CE 0086, 21 CFR (USA) and European Directives compliant for food applications, standards EN420 (General requirements), EN388 (Mechanical Hazards), EN374 (Chemical and Micro-organisms Hazards), EN421 (radioactive contamination only) by the notified laboratory BSI, Manufactured under EC quality assurance, system carried out by the BSI body
Classification PPE Category III
Coating Material Polymer
Color Blue
Disposable Single-use
Exterior Finish Partially Textured
Hand Ambidextrous
Material Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Medium
Sterility Non-sterile
Thickness (Metric) 0.26 mm
Finger Style Textured
For Use With (Application) Short-term laboratory chemical contact, food processing, light duty maintenance, painting, printing, agriculture, electronics, pharmaceutical production
Quantity 50/Pk.
Resistant To Strong and light with good abrasion resistance
Series NT14BPF
Length (Metric) 35 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.