missing translation for 'onlineSavingsMsg'
Learn More

Potassium pyrosulfate, 98%, for analysis

Product Code. 10022640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10022640 500 g Plastic bottle 500g
11914251 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10022640 Supplier Thermo Scientific Acros Supplier No. 207915000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

CAS: 7790-62-7 | K2O7S2 | 254.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-62-7
Molecular Formula K2O7S2
Molecular Weight (g/mol) 254.31
InChI Key KAQHZJVQFBJKCK-UHFFFAOYSA-L
Synonym potassium pyrosulfate, dipotassium disulfate, potassium disulfate, unii-avj6zst7l6, avj6zst7l6, disulfuric acid dipotassium salt, dipotassium sulfonato sulfate, dipotassium disulphate, dipotassium pyrosulfate, kaliumpyrosulfat
PubChem CID 62681
IUPAC Name dipotassium;sulfonato sulfate
SMILES [O-]S(=O)(=O)OS(=O)(=O)[O-].[K+].[K+]

Specifications

Melting Point 325.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 500 g
Assay Percent Range 98%
Linear Formula K2S2O7
Merck Index 15,7784
Solubility Information Solubility in water: may decompose
Formula Weight 254.31
Percent Purity 98%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 2.51
Density 2.5100g/mL
Chemical Name or Material Potassium pyrosulfate
Show More Show Less
compliance-icons
Product Identifier
  • Potassium pyrosulfate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.