missing translation for 'onlineSavingsMsg'
Learn More

Melford Potassium Phosphate, Monobasic

Product Code. 30018709
Click to view available options
Quantity:
1 kg
2.5 kg
Unit Size:
1kg
2.5kg
This item is not returnable. View return policy

Product Code. 30018709

Brand: Melford P412001000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potassium Phosphate, Monobasic, ACS Grade is a chemical compound that holds significance in the life science research, particularly in molecular biology, cell culture, and biochemistry fields.

It is a high-purity reagent with ACS (American Chemical Society) grade certification

Potassium Phosphate, Monobasic, ACS Grade is primarily utilized as a component in laboratory solutions and buffers. It is commonly used as a source of potassium ions and as a pH regulator in various experimental procedures. It is particularly effective in applications requiring slightly acidic to neutral pH conditions. Also, It can be used in protein purification, enzyme assays, nucleic acid research, and other biochemical and molecular biology techniques.

Specifications

Chemical Name or Material Potassium Phosphate, Monobasic, ACS Grade
CAS 7778-77-0
Quantity 1 kg
Molecular Formula KH2PO4
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.