missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate, dibasic, 98+%, ACS reagent

Product Code. 10184333 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic Drum
Plastic bottle
Unit Size:
10kg
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10184333 2.5 kg Plastic bottle 2.5kg
10546982 5 g Glass Bottle 5g
10331211 500 g Plastic bottle 500g
13438578 10 kg Plastic Drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10184333 Supplier Thermo Scientific Acros Supplier No. 424190025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 7758-11-4 | HK2O4P | 174.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11-4-7758
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.18
MDL Number MFCD00011383
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
PubChem CID 24450
ChEBI CHEBI:32031
IUPAC Name dipotassium;hydrogen phosphate
SMILES OP(=O)([O-])[O-].[K+].[K+]

Specifications

Melting Point 340.0°C
Color White
pH 8.5 to 9.6 (5% soln. at 25°C)
Physical Form Fine Crystalline Powder
Quantity 2.5 kg
Assay Percent Range 98+%
Linear Formula K2HPO4
Merck Index 15,7779
Solubility Information Solubility in water: 1600g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 174.18
Percent Purity ≥98%
Grade ACS Reagent
Loss on Drying 1.0% max. (105°C)
Packaging Plastic bottle
Chemical Name or Material Potassium phosphate, dibasic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.