missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Oxidation reagent
Specifications
Specifications
| Melting Point | 350.0°C |
| Color | White |
| pH | 5 to 9 (5% soln. at 20°C) |
| Physical Form | Crystalline Powder |
| Quantity | 250 g |
| Assay Percent Range | 99.5% min. (Iodometry) |
| Linear Formula | KBrO3 |
| Fieser | 11,431 |
| Merck Index | 15, 7738 |
| Solubility Information | Solubility in water: 70g/L (20°C). Other solubilities: practically insoluble in alcohol, soluble in boiling water |
| Show More |
Safety and Handling
Product Identifier
- Potassium bromate
- Danger
- Acute toxicity Category 3
- Carcinogenicity Category 1B
- Oxidising solid Category 1
- H271-May cause fire or explosion; strong oxidiser.
- H301-Toxic if swallowed.
- H350-May cause cancer.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contain: Potassium bromate
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?