Learn More
Greiner Bio-One Polystyrene 384-well Small Volume Microplates
GREENER_CHOICE
Product Code. 10023331
Change view
Click to view available options
Color:
Black
Unit Size:
Pack of 40
Product Code. Color unitSize
10023331 Black Pack of 40
1 options
Product Code. 10023331 Supplier Greiner Bio-One Supplier No. 784076
Only null left
Add to basket
Add to basket
Microplates–Improved detection limit using classical microplate readers. Greiner Bio-One™ Polystyrene 384-Well Small Volume optimizes the Z-axis according to the liquid height to be measured in the wells.
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Traceable to production through a defined LOT number system
  • Ultra low non-specific biomolecular binding properties (proteins, DNA, RNA)
  • Long-term surface stability without degradation or leaching
  • Higher assay sensitivity with reduced background
  • Solid bottom
  • Well profile: Small Volume™
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance DIN ISO 9001
Certified Yes
Format Standard
Lid Without Lid
Array 16 x 24
Packaging Bags of 10
Color Black
Product Type 384-Well Microplate
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 4 to 25 μL
Sterility Non-sterile
Total Volume (Metric) Well 28 μL
Dnase Free Yes
No. per Pack 40Pack
Quantity 40/Cs.
RNase Free Yes