Description
- Adapter has a threaded polypropylene screw cap on top with an inside O-ring for depth adjustment of 6-7mm for straight stem thermometers, glass tubing and other items
- Rotating the extracting nut removes the adapter from flask
- Not autoclavable
Specifications
Specifications
| Depth (Metric) | 6 to 7 mm |
| For Use With (Application) | For 14/20 Tapered Joints |
| Includes | Screw cap with inside O-ring |
| Material | Polypropylene, PTFE |
| Product Type | Thermometer Joint Adapter |
| For Use With (Equipment) | Straight Stem Thermometers, Glass Tubing |