missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Polypropylene Straight Connectors with Conical Nozzles

Product Code. 10138161 Shop All Bürkle Products
Change view
Click to view available options
For Use With:
11 to 14 mm Hose
13 to 15 mm Hose
3 to 5 mm Hose
5 to 7 mm Hose
7 to 10 mm Hose
9 to 12 mm Hose
Unit Size:
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
10138161 11 to 14 mm Hose Pack of 10
10249391 3 to 5 mm Hose Pack of 10
10620143 5 to 7 mm Hose Pack of 10
10198251 7 to 10 mm Hose Pack of 10
10289341 9 to 12 mm Hose Pack of 10
10486891 13 to 15 mm Hose Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10138161 Supplier Buerkle™ Supplier No. 87001214
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured from polypropylene for high thermal resistance. Buerkle™ Polypropylene Straight Connectors with Conical Nozzles resist temperatures up to 135°C.

Cylindrical straight hose connector with conical nozzles
  • Polypropylene
  • Autoclavable to 121°C
  • Suitable for foodstuff
  • Available in a wide range of diameters for varying hose sizes
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EU Foodstuffs, RoHS
Autoclavable Autoclavable
Material Polypropylene
For Use With 11 to 14 mm Hose
Packaging Quantity 10/Pk.; 10/Scale
Shape Straight
Color White
Product Type Hose Tubing Connector
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.