Learn More
Hirschmann™ Polypropylene Microtitration Plate
Description
Not treated disposable plastic with loose glass inserts clear 250μL 384 well Hirschmann
Specifications
Specifications
| Format | Standard |
| Array | 16 x 24 |
| Material | Polypropylene |
| No. of Wells | 384 |
| Total Volume (Metric) Well | 250 μL |
| Includes | Translucent plastic cover included in standard scope of delivery. |