Learn More
Hirschmann™ Polypropylene Disposable Microplate
Product Code. 10182621
Change view
Click to view available options
No. of Wells:
96
Unit Size:
Each
Product Code. No. of Wells unitSize
10182621 96 Each
1 options
Product Code. 10182621 Supplier Hirschmann™ Supplier No. 9240196
Only null left
Add to basket
Add to basket
Disposable plastic plates with fixed glass inserts
Flat bottom not treated disposable plastic with fixed glass inserts clear 1200μL 96 well Hirschmann
Specifications
Format Standard
Array 8 x 12
Color Clear
Disposable Single-use
Product Type Microplate
Material Polypropylene
No. of Wells 96
Total Volume (Metric) Well 1200 μL
Well Shape Flat Bottom (F-well)
No. per Pack 1 Each
Quantity Each