missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poulten & Graf GmbH Polypropylene Adaptor for FORTUNA™ Dispenser with Ground Joint
Description
- Suitable for OPTIFIX and POLYFIX dispenser
- Made of Polypropylene
- Adapter with ground joint
- Can be used for most chemicals and liquids
Specifications
Specifications
| Product Type | Adapters for OPTIFIX and POLYFIX Dispensers |
| Autoclavable | Autoclavable |
| Color | White |
| Description | Adapters for OPTIFIX and POLYFIX dispensers |
| Material | Polypropylene |
| For Use With (Equipment) | OPTIFIX and POLYFIX dispensers |
| Quantity | 1 |
| Thread Size | TS 29 |
| Thread Type | Ground joint |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?