missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Polypropylene Adapter Support

Product Code. 10113402 Shop All Brand Products
Change view
Click to view available options
Color:
Dark Blue
Green
Magenta
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10113402 Green Each
10235032 Magenta Each
10225032 Dark Blue Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10113402 Supplier BRAND™ Supplier No. 26654
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Polypropylene Adapter Support designed for use with accu-jet™ pro.

  • Constructed of polypropylene
  • For use with accu-jet™
TRUSTED_SUSTAINABILITY

Specifications

Color Green
Material Polypropylene
Type Adapter Support
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.