Learn More
Vitlab™ Polymethylpentene Forceps
Product Code. 11717202
Change view
Click to view available options
Length (Metric):
115 mm
145 mm
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
11717202 115 mm Pack of 10
15751996 145 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11717202 Supplier Vitlab™ Supplier No. 67895
Only null left
Add to basket
Edge
Add to basket
Manufactured with PMP for very good resilience. VITLab™ Polymethylpentene Forceps are ideal for use in bottling and decanting.
  • White, Pointed, Elastic 
Specifications
Tip Style Pointed
Material PMP
Length (Metric) 115 mm
Product Type Forceps
Quantity (No. per Case) 10/Pack
For Use With (Application) For autoclaving and microwaves, bottling and decanting
Type Forceps