missing translation for 'onlineSavingsMsg'
Learn More

Ratiolab™ Polyethylene Cleanibag System

Product Code. 10121173
Change view
Click to view available options
Dimensions (L x W):
300 x 200 mm
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
10121173 300 x 200 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10121173 Supplier Ratiolab™ Supplier No. 7000250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Material Polyethylene
Quantity 1000/Pk.
Dimensions (L x W) 300 x 200 mm
Product Type Autoclavable Bags
Capacity (Metric) 1.5 L
Width (Metric) 200 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.