missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(2-acrylamido-2-methyl-1-propanesulfonic acid), 10 wt% aq.sol.; ca. MW 800,000
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Viscous Liquid |
| Linear Formula | {-CH2CH[CONHC(CH3)2CH2SO3H]-}n |
| Quantity | 25 g |
| Percent Purity | 8.0 to 12.0% (solids) |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Chemical Name or Material | Poly(2-acrylamido-2-methyl-1-propanesulfonic acid), 10 wt% aqueous solution |
Safety and Handling
Product Identifier
- Poly(2-acrylamido-2-methyl-1-propanesulfonic acid), 10 wt% solution in water
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?