missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Karl Hecht™ Plexiglas™ Rack
Description
- Holds up to 12 microslides
- Holds slides vertically
- Microscopy and staining accessory
Specifications
Specifications
| Slide Capacity | 12 Microslides |
| Color | White |
| Device Holds | Up to 12 microslides |
| For Use With (Application) | Microscopy and staining |
| For Use With (Equipment) | Microscopy and staining |
| Material | Plexiglass |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?