missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ pLenti7.3/V5-DEST™ Gateway™ Vector Kit

Product Code. 10083844
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10083844 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10083844 Supplier Invitrogen™ Supplier No. V53406

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

The pLenti7.3/V5-DEST™ Gateway™ Vector Kit contains the Gateway™-adapted ViraPower™ HiPerform™ lentiviral expression vector, pLenti7.3/V5-DEST™ for easy recombination-based cloning and high-level expression of a target gene in dividing and non-dividing mammalian cells.

The pLenti7.3/V5-DEST™ Gateway™ Vector Kit contains the Gateway™-adapted ViraPower™ HiPerform™ lentiviral expression vector, pLenti7.3/V5-DEST™ for easy recombination-based cloning and high-level expression of a target gene in dividing and non-dividing mammalian cells. The pLenti7.3/V5-DEST™ vector is equipped with two key genetic elements, making it a HiPerform™ vector: the Woodchuck Posttranscriptional Regulatory Element (WPRE) and the central Polypurine Tract (cPPT) sequence from the HIV-1 integrase gene to produce at least 4-fold increase in protein expression compared to vectors lacking these elements. In addition, the vector allows for an accurate determination of titer of functional lentivirus in just two days using Emerald Green Fluorescent Protein (EmGFP).

Advantages

  • Transient expression
  • Short-term experiments
  • High-throughput screening
  • 2 day titer of functional virus using EmGFP
  • Flexible and versatile Gateway™ recombination cloning technology

Key Features

  • WPRE (Woodchuck Posttranscriptional Regulatory Element) from the woodchuck hepatitis virus, allows increased transgene expression and cPPT (Polypurine Tract) from the HIV-1 integrase gene, increases the copy number of lentivirus integrating into the host genome
  • Human cytomegalovirus (CMV) immediate early promoter to control high-level expression of the gene of interest
  • SV40 promoter driving expression of EmGFP
  • Emerald Green Fluorescent Protein (EmGFP) allows easy determination of Lentiviral titers by flow cytometry and monitor transduced cells.

Kit Includes

  • pLenti7.3/V5-DEST™ Gateway™ Vector box
  • One Shot™ Stbl3™ chemically competent E. coli (Cat. No. C7373-03)

Related SKUs

  • 293FT Cell Line (Cat. No. R70007)
  • ViraPower™ Lentiviral Support Kit (K4970-00)
  • ViraPower™ Lentiviral Packaging Mix (K4975-00)
  • Lipofectamine™ 2000 (11668-019, 11668-027)
TRUSTED_SUSTAINABILITY

Specifications

Constitutive or Inducible System Constitutive
Delivery Type Lentiviral
Format Liquid
Description pLenti7.3/V5-DEST™ Gateway™ Vector Kit, 1 Kit, Liquid, CMV Promoter, V5 Epitope Tag, GFP Reporter, Lentiviral Delivery Method, Gateway™ Cloning Method, pLenti Vector Type, Constitutive System, Used in Cellular Imaging, Protein Assays and Analysis, Protein Expression
Promoter CMV
Product Type Lentiviral Expression Vector
Reporter Gene GFP (EmGFP)
Selection Agent (Eukaryotic) None
Content And Storage pLenti7.3/V5-DEST™ Gateway™ Vector Kit
• pLenti7.3/V5-DEST™ Gateway™ Vector (150 ng/μL), 40 μL
• pLenti7.3/V5-GW/lacZ Control Vector (0.5 μg/μL), 20 μL
Store at -20°C.

One Shot™ Stbl3™ Chemically Competent E. coli box
Transformation reagents including:
• One Shot™ Competent Cells, 20 x 50 μL single-use aliquots
• S.O.C. medium
• pUC19 supercoiled plasmid control
Store at -80°C.
Protein Tag V5 Epitope Tag
Cloning Method Gateway
Quantity 1 Kit
Vector pLenti
Product Line ViraPower, Gateway, HIPERFORM
For Use With (Application) Reporter Assays, Viral Expression
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.