Learn More
Kartell™ Plastilab™ Base and Rod for Vertical Pipette Stand
Custom-designed dish base unit only. Two-tiered pipette holder is not included.
Brand:  Kartell™ 0026200
Product Code. 10142661
  • £144.00 / Pack of 10
Only null left
Estimated Shipment: 17-04-2026
Add to basket
Description
Description
  • Made from polypropylene
  • Small holes in base to allow drainage
  • Boss head allows holder to be raised or lowered on support rod