missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Plastic Stirring Rod

Product Code. 10324982
Change view
Click to view available options
Length (Metric):
130 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
10324982 130 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10324982 Supplier Lovibond™ Supplier No. 364100
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Lovibond Plastic Stirring Rod, Length: 130mm, Quantity: 1/Ea.

Specifications

Length (Metric) 130 mm
Material Plastic
Quantity 1/Ea.
Type Stirring Rod
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.