missing translation for 'onlineSavingsMsg'
Learn More
Learn More
De Buyer™ Plastic Scrapers
Click to view available options
Quantity:
Each
Unit Size:
Each
Description
Order Info
Shape
Specifications
Specifications
| Product Type | Scraper |
| Material | Plastic |
| For Use With (Application) | For corners (scraping), cutting dough or for removing a semi-compact to compact preparation from a container, for filling pastry piping bags. |
| Length (Metric) | 150 mm |
| Quantity | Each |
| Shape | Flat and Rounded Edges |
| Width (Metric) | 110 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction