missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hirschmann™ Plastic Base and Safety Ring
Description
- For measuring cylinders
- Without base
- Available to fit a range of sizes
Specifications
Specifications
| Color | Green |
| Capacity (Metric) | 10 mL |
| For Use With (Equipment) | Measuring Cylinder |
| No. per Pack | 1 Each |
| Includes | Protecting Ring |
| Type | Base |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?