Learn More
ratiolab™ Pipet Tips
Product Code. 10432311
Change view
Click to view available options
Color:
Blue
Clear
Yellow
Sterility:
Non-sterile
Sterile
Volume (Metric):
0.1 to 10 μL
0.5 to 20 μL
1 to 200 μL
1 to 250 μL
100 to 1000 μL
100 to 1200 μL
500 to 2500 μL
Packaging:
Bag
Multirack
Unit Size:
Pack of 1000
Pack of 250
Pack of 600
Pack of 960
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Sterility Volume (Metric) Packaging unitSize
10432311 Clear Non-sterile 0.1 to 10 μL Bag Pack of 1000
15110386 Clear Sterile 0.5 to 20 μL Multirack Pack of 960
12366412 Clear Sterile 0.1 to 10 μL Multirack Pack of 960
15472580 Clear Non-sterile 500 to 2500 μL Bag Pack of 250
10737294 Clear Non-sterile 1 to 200 μL Bag Pack of 1000
11303310 Clear Non-sterile 1 to 250 μL Bag Pack of 1000
10140843 Clear Non-sterile 100 to 1000 μL Bag Pack of 1000
10564605 Blue Sterile 100 to 1200 μL Multirack Pack of 600
10458504 Blue Non-sterile 100 to 1200 μL Multirack Pack of 600
12344673 Yellow Sterile 1 to 200 μL Multirack Pack of 960
16224822 Yellow Non-sterile 1 to 250 μL Multirack Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 10432311 Supplier ratiolab™ Supplier No. 2500190
Only null left
Add to basket
Add to basket
Pipet tips for all commercially available manual or electronic, single or multichannel pipets.
  • Guaranteed dust-free due to special ionization treatment
  • High transparency due to high quality surfaces
  • Minimal residual wetting due to the controlled quality of raw materials
  • High volume accuracy due to precise tip orifice
  • Together with specially developed Multirack, these make pipetting much simpler and safer in the most sensitive applications
Specifications
Volume (Metric) 0.1 to 10 μL
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Packaging Bag
Quantity 1000 Tips
Material Polypropylene