missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PIM3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 10098929
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10098929 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10098929 Supplier Invitrogen™ Supplier No. PA513976

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is predicted to react with rat based on sequence homology.

PIM3 serine/threonine kinase or PIM-3 oncogene belongs to a family of protooncogenes. PIM3 is expressed in hepatocellular carcinoma development and plays a main role in the proliferation of human hepatoma cell lines. PIM3 also expressed in human pancreatic cancer cells and phosphorylates BAD to block Bad-mediated apoptosis in human pancreatic cancer cell lines. PIM3 is widely expressed, with the exception of no expression observed in colon, thymus, and small intestine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIM3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PIM3
Gene Accession No. P58750, Q86V86
Gene Alias BC026639; Kid1; KID-1/kinase induced by depolarization; Pim3; pim-3; pim-3 oncogene; Pim-3 proto-oncogene, serine/threonine kinase; proviral integration site 3; serine/threonine kinase Pim-3; serine/threonine-protein kinase pim-3
Gene Symbols PIM3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 259-288 amino acids from the C-terminal region of human PIM3.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223775, 415116
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.