Learn More
Ansell Picolon Confort Gloves
Product Code. 17774879
Change view
Click to view available options
Size:
7
10
Unit Size:
12 pairs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17774879 10 12 pairs
17764879 7 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17774879 Supplier Ansell Supplier No. C357100
Only null left
Add to basket
Add to basket
Picolon Confort Gloves are non-stop seamless knitted gloves.
  • Stretchable seamless liner for great fit and dexterity
  • Dots on palm and fingers for enhanced grip properties
  • Good resistance to abrasion and small blade cut hazards
  • Lint free glove
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) CE Number, Ecovadis label, EN 388:2016 +A1:2018, EN ISO 21420:2020, Category II, REACH Compliant
Chlorinated No
Coated Area Palm
Coating Material PVC Dots
Color White
Exterior Finish Partially Textured
Hand Specific
Latex Free Yes
Material PVC
Product Type PVC-coated Industrial Glove
Size 10
Glove Cuff Style Knit
Liner Material Polyamide
Resistant To resistance to abrasion and low, blade cut hazards, in addition to being lint-free