missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-PAX2 (Ser393) Polyclonal Antibody
GREENER_CHOICE

Product Code. 10058382
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10058382 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10058382 Supplier Invitrogen™ Supplier No. 369200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Rabbit Polyclonal Antibody

Phospho-PAX2 (Ser393) Polyclonal Antibody for Western Blot, IHC (P), ELISA

The pax genes are a family of transcription factors that are active in specific tissues during early embryonic development. Pax family members possess a DNA-binding domain encoded by the paired box. Because paired domain containing genes encode transcription factors, they are capable of executing a genetic program. Several pax genes have also been associated with developmental mutations including: pax-3, which is associated with Waardenburg syndrome, pax-6, which is associated with Aniridia, and pax-2, which is associated with Wilms tumor. During embryogenesis, Pax-2 is expressed in the developing kidney. In particular, the pax-2 gene is expressed in condensing metanephric mesenchyme and in early epithelial structures derived from mesenchyme; however, pax-2 mRNA and protein levels are rapidly down regulated as the tubular epithelium matures. Although Pax-2 is down regulated during renal epithelium maturation, Pax-2 expression persists in the undifferentiated epithelium of Wilms' tumors. Persistent expression of Pax-2 in Wilms' tumors occurs frequently and correlates with the proliferation of poorly differentiated epithelial cells in these tumors. Interestingly, expression of the Wilms' tumor suppresser protein, WT1, coincides with down-regulation of the pax-2 gene; thus, suggesting that WT1 can directly repress pax-2 transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PAX2 (Ser393)
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene PAX2
Gene Accession No. P32114, Q02962
Gene Alias FSGS7; Opdc; optic disc coloboma; paired box 2; paired box gene 2; paired box homeotic gene 2; paired box protein Pax-2; PAPRS; PAX2; Pax-2
Gene Symbols PAX2
Host Species Rabbit
Immunogen Synthetic phospho-peptide derived from the sequence surrounding the phosphorylated Ser393 residue of mouse and human Pax2 (paired box protein 2). Ser393 of the 415 amino acid mouse Pax2 isoform is equivalent to Ser394 of the 416 amino acid human Pax2 isoform, and to Ser371 of the 393 amino acid human Pax2 isoform.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18504, 293992, 5076
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.