missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-Connexin 43 (Ser368) Polyclonal Antibody
GREENER_CHOICE

Product Code. 10174543
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10174543 100 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10174543 Supplier Invitrogen™ Supplier No. 483000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Rabbit Polyclonal Antibody

This antibody is predicted to work in Bovine, Canine, Chicken, Guinea Pig, Human, Zebrafish due to the sequence used as antigen being 100% homologous.

Connexin 43 (Cx43) is a member of the gap junction protein family. Connexins assemble as a hexamer and are transported to the plasma membrane to create a hemichannel that can associate with hemichannels on nearby cells to create cell-to-cell channels. Clusters of these channels assemble to make gap junctions. Gap junction communication is important in development and regulation of cell growth. Phosphorylation of Cx43 is important in regulating assembly and function of gap junctions. Ser368 of Cx43 is phosphorylated by protein kinase C (PKC) after activation by phorbol esters, which decreases cell-to-cell communication. Src can interact with and phosphorylate Cx43 to alter gap junction communication. GFAP are membrane-spanning proteins that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Connexin 43 is the major protein of gap junctions in the heart, and gap junctions are thought to have a crucial role in the synchronized contraction of the heart and in embryonic development. Connexin 43 is also targeted by several protein kinases that regulate myocardial cell-cell coupling. A related intron-less connexin 43 pseudogene, GJA1P, has been mapped to chromosome 5. Mutations in the GFAP gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy, oculodentodigital dysplasia and heart malformations. Alternatively spliced transcript variants of GFAP have been found.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Connexin 43 (Ser368)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 0.15M NaCl, 100 μg/mL BSA, 50% glycerol and no preservative; pH 7.5
Gene GJA1
Gene Accession No. P08050, P23242
Gene Alias alpha 1 connexin; AU042049; AVSD3; AW546267; CMDR; Cnx43; connexin 43; connexin43; connexin-43; cx43; Cx-43; cx43 protein; cx43a1; Cx43alpha1; CXA1; Cxn-43; DFNB38; EKVP; etID309742.9; gap junction 43 kDa heart protein; Gap junction alpha-1 protein; gap junction membrane channel protein; gap junction membrane channel protein alpha 1; gap junction protein; gap junction protein alpha 1; gap junction protein connexin 43; gap junction protein, alpha 1; gap junction protein, alpha 1, 43 kD (connexin 43); gap junction protein, alpha 1, 43kDa; gap junction protein, alpha 1, 43kDa (connexin 43); gja1; Gja-1; GJAL; HLHS1; HSS; Npm1; ODD; ODDD; ODOD; PPKCA; SDTY3; shf; Short fin protein; sof; stopsel; stp; Syndactyly type III; vascular smooth muscle connexin43; Vascular smooth muscle connexin-43; zfCx43.3
Gene Symbols GJA1
Host Species Rabbit
Immunogen Phosphopeptide corresponding to amino acid residues surrounding the phosphoserine 368 of rat connexin 43.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14609, 24392
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.