missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Phosphate Ortho/Total Cuvette Tests

Product Code. 10014311
Change view
Click to view available options
Range:
0.5 to 5mg/L PO4-P
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range unitSize
10014311 0.5 to 5mg/L PO4-P Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10014311 Supplier Hach Lange™ Supplier No. LCK348

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pre-dosed reagents for maximum safety. Easy to use and photometric evaluation, packaged in a fully equipped box. Officially approved for consent limits. Environmentally friendly due to less chemicals.

  • Phosphate (Ortho/Total) cuvette test 0.05-1.5 mg/L PO4-P
  • For use with DR3900, DR6000, DR1900, DR2800, DR3800, DR5000 instruments
  • ISO 6878-1-1986, DIN 38405 D11-4 certifications
  • Shelf life of 23 months from production date
  • Storage conditions of 15 - 25 °C
  • Measuring ranges of 0.05 - 1.5 mg/L PO4-P and 0.15 - 4.5 mg/L PO4
  • Precise and reliable measurement values
  • Convenient and error-free dosing of the reagents
TRUSTED_SUSTAINABILITY

Specifications

No. of Tests 25
Range 0.5 to 5mg/L PO4-P
Storage Requirements 15°C to 25°C
Test Method Phosphormolybdenum Blue
Type Phosphate Ortho/Total Cuvette Test
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phosphat/Phosphate, DosiCapZip
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H302-Harmful if swallowed.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
Supplemental information
  • MIXTURE LIST-Contain: Sodium peroxidisulfate, Sodium metaborate, Lithium sulfate monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.