missing translation for 'onlineSavingsMsg'
Learn More

Phenol Detached Crystals, Certified AR, for Analysis, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™

Product Code. p-7073387 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2 kg
Unit Size:
1kg
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10734381 500 g 500g
10569380 1 kg 1kg
10345810 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10734381 Supplier Fisher Chemical Supplier No. P/2360/53
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

C6H6O, CAS Number-108-95-2, oxybenzene, hydroxybenzene, carbolic acid, phenyl hydroxide, monophenol, phenyl hydrate, phenic acid, phenylic acid, phenylic alcohol, benzenol, 1kg, 182 deg.C, CHEBI:15882, Colorless to White, 94.11g/mol, ISWSIDIOOBJBQZ-UHFFFAOYSA-N, 39 deg.C, 2143, 94.113 | CAS: 108-95-2 | C6H6O | 94.11 g/mol

C6H6O, CAS Number-108-95-2, oxybenzene, hydroxybenzene, carbolic acid, phenyl hydroxide, monophenol, phenyl hydrate, phenic acid, phenylic acid, phenylic alcohol, benzenol, 1kg, 182 deg.C, CHEBI:15882, Colorless to White, 94.11g/mol, ISWSIDIOOBJBQZ-UHFFFAOYSA-N, 39 deg.C, 2143, 94.113

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

Melting Point 39°C
Color Colorless to White
pH 6
Boiling Point 182°C
Packaging Plastic powder jar
Quantity 500 g
Viscosity 3.437 mPaS (50°C)
Formula Weight 94.11g/mol
Vapor Pressure 0.4mbar at 20°C
Physical Form Crystalline Solid
Chemical Name or Material Phenol Detached Crystals
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.