missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Melting Point | 40.8°C |
| Boiling Point | 181.8°C (1013 hPa) |
| CAS | 108-95-2 |
| Clarity | Passes Test |
| Assay Percent Range | min. 99.0% |
| Molecular Formula | C6H6O |
| MDL Number | MFCD00002143 |
| Synonym | Carbolic acid , Hydroxybenzene , Phenylalcohol |
| InChI Key | ISWSIDIOOBJBQZ-UHFFFAOYSA-N |
| SMILES | OC1=CC=CC=C1 |
| Show More |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 3
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H341-Suspected of causing genetic defects.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?