missing translation for 'onlineSavingsMsg'
Learn More

Phenanthrene, 97%

Product Code. 10183320 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
500g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10183320 500 g Plastic bottle 500g
10745241 5 g Glass bottle 5g
10205040 50 g Plastic bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10183320 Supplier Thermo Scientific Acros Supplier No. 130095000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 85-01-8 | C14H10 | 178.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-01-8
Molecular Formula C14H10
Molecular Weight (g/mol) 178.23
MDL Number MFCD00001168
InChI Key YNPNZTXNASCQKK-UHFFFAOYSA-N
Synonym phenanthren, phenanthrin, phenanthracene, ravatite, phenantrin, phenanthren german, phenanthrene, pure, unii-448j8e5bst, ccris 1233, chembl46730
PubChem CID 995
ChEBI CHEBI:28851
IUPAC Name phenanthrene
SMILES C1=CC=C2C(C=CC3=CC=CC=C23)=C1

Specifications

Melting Point 95°C to 101°C
Color White to Gray
Density 1.0630g/mL
Boiling Point 336°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Quantity 500 g
Beilstein 05, 667
Merck Index 15, 7326
Specific Gravity 1.063
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetic acid,soluble in alcohol,ether,benzene and carbon,disulfide
Formula Weight 178.23
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Phenanthrene, 98+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.