missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 12.00, Chem-Lab
Description
pH Levels ± 0.02:
- pH 12.58 — 0°C
- pH 12.26 — 10°C
- pH 12.00 — 20°C
- pH 11.88 — 25°C
- pH 11.75 — 30°C
- pH 11.53 — 40°C
- pH 11.31 — 50°C
- pH 11.09 — 60°C
- pH 10.88 — 70°C
- pH 10.68 — 80°C
- pH 10.48 — 90°C
Specifications
Specifications
| CAS | 14265-44-2, 1310-73-2 |
| Melting Point | 0°C |
| Boiling Point | 100°C |
| Color | Colorless |
| Density | 1.01g/mL |
| For Use With (Application) | Laboratory use, pH measurements |
| pH | 12.00 ±0.02 |
| Physical Form | Liquid |
| Quantity | 1000 mL |
| Solution Type | pH Buffer |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?