missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PFKFB3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 10121030
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10121030 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10121030 Supplier Invitrogen™ Supplier No. PA515456

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is predicted to react with rat based on sequence homology.

The protein encoded by this gene belongs to a family of bifunctional proteins that are involved in both the synthesis and degradation of fructose-2,6-bisphosphate, a regulatory molecule that controls glycolysis in eukaryotes. The encoded protein has a 6-phosphofructo-2-kinase activity that catalyzes the synthesis of fructose-2,6-bisphosphate (F2,6BP), and a fructose-2,6-biphosphatase activity that catalyzes the degradation of F2,6BP. This protein is required for cell cycle progression and prevention of apoptosis. It functions as a regulator of cyclin-dependent kinase 1, linking glucose metabolism to cell proliferation and survival in tumor cells. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PFKFB3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PFKFB3
Gene Accession No. Q16875
Gene Alias 6-biphosphatase 3; 6PF-2-K/Fru-2,6-P2ase 3; 6PF-2-K/Fru-2,6-P2ase brain/placenta-type isozyme; 6PF-2-K/Fru-2,6-P2ASE brain-type isozyme; 6-phosphofructo-2-kinase; 6-phosphofructo-2-kinase/ fructose-2,6-bisphosphatase; 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3; 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 3; E330010H22Rik; Fructose-2; Fructose-2,6-bisphosphatase; fructose-6-phosphate,2-kinase/fructose-2, 6-bisphosphatase; inducible 6-phosphofructo-2-kinase; inducible 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase; IPFK2; iPFK-2; PFK/FBPase 3; PFK2; PFKFB3; RB2K; renal carcinoma antigen NY-REN-56; RP11-298K24.3; uPFK-2
Gene Symbols PFKFB3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 454-484 amino acids from the C-terminal region of human PFKFB3.
Purification Method Ammonium Sulfate Precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5209
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.