missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Perlecan Monoclonal Antibody (7B5)
GREENER_CHOICE

Product Code. 10109352
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10109352 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10109352 Supplier Invitrogen™ Supplier No. 134400

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Mouse Monoclonal Antibody

Perlecan Monoclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot

Perlecan is a major heparan-sulfate proteoglycan (HSPG) found within all basement membranes and cell surfaces. Because of its strategic location and ability to store and protect growth factors, perlecan has been strongly implicated in the control of tumor cell growth and metastatic behavior. Perlecan possesses angiogenic and growth-promoting attributes primarily by acting as a coreceptor for basic fibroblast growth factor (FGF-2). Suppression of perlecan causes substantial inhibition of neoplastic growth and neovascularization. Thus, perlecan is a potent inducer of neoplasm growth and angiogenesis in vivo and therapeutic interventions targeting this key modulator of tumor progression may improve neoplastic treatment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Perlecan
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 7B5
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene HSPG2
Gene Accession No. P98160
Gene Alias AI852380; Basement membrane-specific heparan sulfate proteoglycan core protein; Endorepellin; endorepellin (domain V region); heparan sulfate proteoglycan 2; HSPG; Hspg2; LG3 peptide; LOC313641; Pcn; per; perlecan; perlecan (heparan sulfate proteoglycan 2); perlecan proteoglycan; PLC; PRCAN; RP11-132G19.2; SJA; SJS; SJS1
Gene Symbols HSPG2
Host Species Mouse
Immunogen rec-Perlecan domain III.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3339
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.