missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PCAF Monoclonal Antibody (E.107.0)
GREENER_CHOICE

Product Code. 10060308
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10060308 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10060308 Supplier Invitrogen™ Supplier No. MA511186

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

It is not recommended to aliquot this antibody. This antibody is not cross-reactive with the related GCN5L2 protein.

PCAF (histone acetyltransferase KAT2B) functions as a histone acetyltransferase (HAT) to promote transcriptional activation. It has significant histone acetyltransferase activity with core histones (H3 and H4) and nucleosome core particles. PCAF also acetylates non-histone proteins such as ACLY, PLK4, and TBX5. It inhibits cell-cycle progression and counteracts the mitogenic activity of the adenoviral oncoprotein E1A. PCAF also acts as a circadian transcriptional coactivator which enhances the activity of the circadian transcriptional activators NPAS2-ARNTL/BMAL1 and CLOCK-ARNTL/BMAL1 heterodimers. It is involved in heart and limb development by mediating acetylation of TBX5 and acetylation regulating nucleocytoplasmic shutting of TBX5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCAF
Applications ChIP Assay, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone E.107.0
Concentration 358 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5
Gene KAT2B
Gene Accession No. Q92831, Q9JHD1
Gene Alias A930006P13Rik; AI461839; AW536563; CAF; CREBBP-associated factor; histone acetylase PCAF; Histone acetyltransferase KAT2B; histone acetyltransferase PCAF; K(lysine) acetyltransferase 2B; Kat2b; Lysine acetyltransferase 2B; P/ PCAF; P/CAF; P300/CBP-associated factor; Pcaf; Spermidine acetyltransferase KAT2B
Gene Symbols KAT2B
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the amino terminus of human PCAF protein.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18519, 301164, 8850
Target Species Human, Mouse, Monkey, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.