missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ CCR5 Activated GPCR Internalization U2OS Cell Line

Product Code. 17343599
Encompass
Change view
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17343599 2 Vials 2 ampoules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17343599 Supplier DiscoverX Supplier No. 930653C3

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PathHunter™ CCR5 Activated GPCR Internalization U2OS Cell Line measures CCR5 (GPCR) internalization via early-endosome localization. This cell line ships as two vials of cryopreserved cells.

The PathHunter™ CCR5 Activated GPCR internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line enables assessment of ligand (e.g. CCL3/MIP-1α) based internalization of CCR5 via detection of localization to the early endosome. This product uses EFC technology. The included cell line overexpresses EA-tagged β-arrestin 2 as well as a PK-tagged early endosome protein and untagged CCR5. Upon ligand-induced receptor activation, β-arrestin 2 is recruited to the activated GPCR and the resulting CCR5-arrestin complex is internalized to the early endosome, leading to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifications

Assay Entry Receptor Internalization
Cell Line U2OS
For Use With (Application) Cell Lines
Primary Agonist Recombinant Human MIP-1α (CCL3)
Quantity 2 Vials
Target Entry CCR5
Accession Number BC038398
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.