missing translation for 'onlineSavingsMsg'
Learn More

Paritaprevir, MedChemExpress

Product Code. 16491467
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491467 50 mg 50mg
16485258 10 mg 10mg
16447977 5 mg 5mg
16435926 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491467 Supplier Medchem Express Supplier No. HY12594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Paritaprevir (ABT-450) is a potent, orally active and antiviral non-structural protein 3/4A (NS3/4A) protease inhibitor with EC50s of 1 and 0.21 nM against HCV 1a and 1b, respectively. Paritaprevir is also a SARS-CoV 3CLpro inhibitor with an IC50 of 1.31 μM. Paritaprevir is metabolized primarily by cytochrome P450 (CYP) 3A. The plasma concentration and half-life of Paritaprevir can be enhanced by Ritonavir (a CYP450 inhibitor).

Paritaprevir (ABT-450) is a potent, orally active and antiviral non-structural protein 3/4A (NS3/4A) protease inhibitor with EC50s of 1 and 0.21 nM against HCV 1a and 1b, respectively. Paritaprevir is also a SARS-CoV 3CLpro inhibitor with an IC50 of 1.31 μM. Paritaprevir is metabolized primarily by cytochrome P450 (CYP) 3A. The plasma concentration and half-life of Paritaprevir can be enhanced by Ritonavir (a CYP450 inhibitor).

Specifications

Chemical Name or Material Paritaprevir
CAS 1216941-48-8
Color White
Purity Grade Notes Research
Molecular Formula C40H43N7O7S
Quantity 50 mg
Synonym ABT-450 Veruprevir
Solubility Information DMSO : ≥ 125 mg/mL (163.21 mM) ∣H2O : ≥ 0.1 mg/mL (0.13 mM)
SMILES O=C([C@]1(NC([C@@]2([H])N3C[C@H](OC4=C(C=CC=C5)C5=C6C(C=CC=C6)=N4)C2)=O)[C@H](/C=C\CCCCC[C@H](NC(C7=NC=C(C)N=C7)=O)C3=O)C1)NS(=O)(C8CC8)=O
Molecular Weight (g/mol) 765.88
Formula Weight 765.88
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.