Product Code. 30426666
Click to view available options
Quantity:
500 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
unitSize
30426666
10 g
10g
30422702
500 mg
500mg
30415832
1 g
-
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code.
30426666
Supplier
Toronto Research Chemicals
Supplier No.
TRCP19140210G
Please sign in to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Chemical Identifiers
CAS
32008-59-6
Molecular Formula
(C 2H2 O)n
Molecular Weight (g/mol)
64.077
Synonym
Poly(oxymethylene-d2) (8CI, 9CI), Paraformaldehyde-d2
SMILES
[X]OC([2H])([2H])OC([2H])([2H])OC([X])([2H])[2H]
Specifications
Quantity
10 g
Formula Weight
64.046
Chemical Name or Material
Paraformaldehyde-d2
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?