missing translation for 'onlineSavingsMsg'
Learn More

Paraformaldehyde-D2, TRC

Product Code. 30422702
Change view
Click to view available options
Quantity:
500 mg
1 g
10 g
Unit Size:
10g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422702 500 mg 500mg
30426666 10 g 10g
30415832 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30422702 Supplier Toronto Research Chemicals Supplier No. TRCP191402500MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32008-59-6
Molecular Formula (C 2H2 O)n
Molecular Weight (g/mol) 64.077
Synonym Poly(oxymethylene-d2) (8CI, 9CI), Paraformaldehyde-d2
SMILES [X]OC([2H])([2H])OC([2H])([2H])OC([X])([2H])[2H]

Specifications

Quantity 500 mg
Formula Weight 64.046
Chemical Name or Material Paraformaldehyde-d2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.