missing translation for 'onlineSavingsMsg'
Learn More

Paraformaldehyde-d2, CDN

Product Code. 30311083
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30311083 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30311083 Supplier CDN Supplier No. CDND01835G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Paraformaldehyde-d2
CAS 32008-59-6
Molecular Formula (C 2H2 O)n
Quantity 5 g
Synonym Poly(oxymethylene-d2) (8 CI, 9 CI), Paraformaldehyde-d2
SMILES [X]OC([2 H])([2 H])OC([2 H])([2 H])OC([X])([2 H])[2 H]
Molecular Weight (g/mol) 64.077
Formula Weight 64.046 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.