missing translation for 'onlineSavingsMsg'
Learn More

BOLA Paddle

Product Code. 10094273
Change view
Click to view available options
Dimensions (L x W x H):
80 x 24 x 4 mm
80 x 25 x 5 mm
110 x 25 x 5 mm
140 x 25 x 5 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
10094273 80 x 25 x 5 mm Each
10769254 80 x 24 x 4 mm Each
10790075 110 x 25 x 5 mm Each
13437679 140 x 25 x 5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10094273 Supplier BOLA Supplier No. C44610
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Paddle for stainless steel shafts jacketed with PTFE, glass shafts or metal shafts

  • These solid stirrer blades are made of PTFE and have a set of clamp screws made of a PTFE/PEEK compound.
  • The blades can be fixed tightly on BOLA Stirrer Shafts by means of the clamp screws.
  • A spanner wrench is included for easy assembly.

Specifications

Diameter (Metric) Shaft 10 mm
Dimensions (L x W x H) 80 x 25 x 5 mm
Coating PTFE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.