missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ p19 INK4d Monoclonal Antibody (2D2G4)
GREENER_CHOICE

Product Code. 10067702
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10067702 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10067702 Supplier Invitrogen™ Supplier No. 378700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Mouse Monoclonal Antibody

p19 INK4d Monoclonal Antibody for Western Blot, IP, ELISA

p19 is a member of the INK4 family of cyclin-dependent kinase inhibitors. This protein has been shown to form a stable complex with CDK4 or CDK6, and prevent the activation of the CDK kinases, thus function as a cell growth regulator that controls cell cycle G1 progression. The abundance of the transcript of this gene was found to oscillate in a cell-cycle dependent manner with the lowest expression at mid G1 and a maximal expression during S phase. The negative regulation of the cell cycle involved in this protein was shown to participate in repressing neuronal proliferation, as well as spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p19 INK4d
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2D2G4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene CDKN2D
Gene Accession No. P55273
Gene Alias CDK inhibitor p19INK4d; CDK4I; CDKN2D; cell cycle inhibitor, Nur77 associating protein; cyclin dependent kinase inhibitor 2D; cyclin-dependent kinase 4 inhibitor D; cyclin-dependent kinase 4 inhibitor D p19; cyclin-dependent kinase inhibitor 2D (p19, inhibits CDK4); inhibitor of cyclin-dependent kinase 4d; INK4d; MTS1; p16-INK4; p16-INK4a; p19; p19INK4d; p19-INK4D; similar to cyclin-dependent kinase inhibitor 2D
Gene Symbols CDKN2D
Host Species Mouse
Immunogen A recombinant full length human p19 protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1032
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.