missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonic acid, pure, 12% in acetic acid

Product Code. 10015720 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10015720 1 kg 1kg
10677391 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10015720 Supplier Thermo Scientific Acros Supplier No. 421220010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104-15-4 | C7H8O3S | 172.198 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material p-Toluenesulfonic acid
Color Colorless to Yellow
Name Note 12% in Acetic Acid
CAS 104-15-4, 64-19-7
Boiling Point 116.0°C
Molecular Formula C7H8O3S
MDL Number MFCD00064387
Beilstein 11, 97
Concentration 11 to 13% (Alkalimetry)
Fieser 01,1172; 04,508; 05,673; 06,597; 07,374; 08,488; 09,471; 11,535; 12,507
Density 1.0700g/mL
Merck Index 15, 9692
Synonym p-toluenesulfonic acid, p-toluenesulphonic acid, 4-toluenesulfonic acid, tosic acid, p-tolylsulfonic acid, toluene-4-sulfonic acid, toluenesulfonic acid, tosylic acid, benzenesulfonic acid, 4-methyl, p-methylbenzenesulfonic acid
Solubility Information Solubility in water: soluble.
InChI Key JOXIMZWYDAKGHI-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)S(=O)(=O)O
IUPAC Name 4-methylbenzenesulfonic acid
Molecular Weight (g/mol) 172.198
PubChem CID 6101
ChEBI CHEBI:27849
Formula Weight 172.2
Grade Pure
Packaging Glass bottle
Refractive Index 1.3825 to 1.3845
Specific Gravity 1.07
Quantity 1 kg
Flash Point 41°C
Physical Form Solution
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • p-Toluenesulfonic acid, 12% in acetic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.