missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Toluene-d7-sulfonic Acid H2O, TRC
Specifications
Specifications
| Chemical Name or Material | p-Toluene-d7-sulfonic Acid H2O |
| CAS | 1219795-22-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C7 D7 H O3 S . H2 O |
| Quantity | 100 mg |
| Synonym | p-Toluene-d7-sulfonic acid hydrate, p-Toluenesulfonic-d7 acid monohydrate, 4-Methylbenzenesulfonic-d7 acid monohydrate, 4-(Methyl-d3)benzenesulfonic-2,3,5,6-d4 acid monohydrate, 4-Methylbenzenesulfonic acid D7 monohydrate |
| InChI Formula | InChI=1S/C7H8O3S.H2O/c1-6-2-4-7(5-3-6)11(8,9)10;/h2-5H,1H3,(H,8,9,10);1H2/i1D3,2D,3D,4D,5D; |
| SMILES | O.[2H]c1c([2H])c(c([2H])c([2H])c1C([2H])([2H])[2H])S(=O)(=O)O |
| IUPAC Name | 2,3,5,6-tetradeuterio-4-(trideuteriomethyl)benzenesulfonic acid;hydrate |
| Molecular Weight (g/mol) | 197.26 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?