missing translation for 'onlineSavingsMsg'
Learn More

Oxyphenisatine, MedChemExpress

Product Code. 16498836
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498836 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16498836 Supplier Medchem Express Supplier No. HYB2102

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Oxyphenisatine (Oxyphenisatin) is a laxative. Oxyphenisatin acetate is the pro-drug of oxyphenisatin with anticancer activity.

Oxyphenisatine (Oxyphenisatin) is a laxative. Oxyphenisatin acetate is the pro-drug of oxyphenisatin with anticancer activity.

Specifications

Chemical Name or Material Oxyphenisatine
CAS 125-13-3
Color White
Purity Grade Notes Research
Molecular Formula C20H15NO3
Quantity 100 mg
Synonym Oxyphenisatin
Solubility Information DMSO : ≥ 100 mg/mL (315.12 mM)
SMILES O=C1NC2=C(C=CC=C2)C1(C3=CC=C(O)C=C3)C4=CC=C(O)C=C4
Molecular Weight (g/mol) 317.34
Formula Weight 317.34
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.