Learn More
Thermo Scientific™ Streptococcal Grouping Kit using Latex Agglutination
Product Code. 10127942
Change view
Click to view available options
Description:
Enzyme Extraction Reagent
Grouping Kit
Polyvalent Positive Control
Reagent A
Reagent B
Reagent C
Reagent D
Reagent F
Reagent G
Unit Size:
50 tests
Pack of 2
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
10127942 Reagent G 50 tests
11479878 Grouping Kit 50 tests
11903522 Reagent A 50 tests
11913522 Reagent B 50 tests
10537283 Reagent C 50 tests
11973932 Reagent D 50 tests
10770354 Reagent F 50 tests
10035812 Polyvalent Positive Control 50 tests
13275599 Enzyme Extraction Reagent Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10127942 Supplier Thermo Scientific™ Supplier No. DR0591G
Rapidly identify streptococcal groups A, B, C, D, F, and G by latex agglutination using this test kit.
Rapidly identify streptococcal groups A, B, C, D, F, and G by latex agglutination using Thermo Scientific™ Oxoid™ Streptococcal Grouping Kit. This rapid latex agglutination test is designed to deliver simple, rapid and sensitive results for the identification of streptococcal groups A, B, C, D, F, and G . The test utilizes latex particles coated with group-specific antibodies which will agglutinate in the presence of homologous antigens.
  • Supplied with rapid latex agglutination test
TRUSTED_SUSTAINABILITY
Specifications
Description Reagent G
Type Reagents for Streptococcus Testing
Organism Group Group A, B, C, D, F and G
Packaging Type 50 Tests/Vial