missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ MUG Supplement

Product Code. 10015972
Change view
Click to view available options
Quantity:
10 x 50 mg
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10015972 10 x 50 mg Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10015972 Supplier Thermo Scientific™ Supplier No. BR0071E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Oxoid MUG Supplement (50mg) Fluorescent agent is used for the detection of E. coli.

Oxoid MUG Supplement (50mg) Fluorescent agent is used for the detection of E. coli.

  • Contains 4-methylumbelliferyl-β-D-glucuronide (MUG)
  • Each vial supplements 500mL of medium
TRUSTED_SUSTAINABILITY

Specifications

Description MUG Supplement
Form Freeze-dried
Product Type Microbiology Media
Quantity 10 x 50 mg
Yield Yield will be dependent upon formulation; follow the guidelines for your specific product.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.