missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MRVP Medium (Dehydrated)

Product Code. 10108102
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10108102 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10108102 Supplier Thermo Scientific™ Supplier No. CM0043B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Differentiate coli-aerogenes group by Methyl-red and Voges-Proskauer test with this glucose phosphate medium.

Differentiate coli-aerogenes group by Methyl-red and Voges-Proskauer tests with Thermo Scientific™ Oxoid™ MRVP Medium (Dehydrated). This glucose-phosphate medium is recommended for the Methyl-red and Voges-Proskauer tests, for the differentiation of the coli-aerogenes group1. Both of these tests are based on the detection of specific breakdown products of carbohydrate metabolism.

Use MRVP Broth for differentiation of coli-aerogenes group by the Methyl-red and Voges-Proskauer test.

Smith2 noted the low acid production of Enterobacter aerogenes cultures as compared with those of Escherichia coli. Clark & Lubs3 employed methyl-red as a hydrogen-ion concentration indicator in order to differentiate glucose phosphate peptone water cultures of members of the coli-typhoid group. This test, now known as the Methyl-red test, distinguishes those organisms able to form large amounts of acid from glucose so that the pH falls below 4.4 and those organisms which cannot produce a low pH level.

The difference in pH value is visualised by adding methyl-red to the culture, (<pH 4.4 red: pH 5.0-5.8 orange: >pH 6.0 yellow).

Voges & Proskauer4 described a red fluorescent coloration that appeared after the addition of potassium hydroxide to cultures of certain organisms in glucose medium. The coloration was shown to be due to the oxidation of the acetylmethyl-carbinol producing diacetyl which reacts with the peptone of the medium to give a red colour5,6. Durham7 noted that Enterobacter aerogenes gave a positive reaction but that Escherichia coli produced no coloration, and it later became clear that there was a negative correlation between the Methyl-red and Voges-Proskauer tests8,9 for lactose-fermenting coliform organisms.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description MRVP Medium (Clarks and Lubs Medium)
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 29.4L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.