missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Malt Extract Broth (Dehydrated)

Product Code. 10031752
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10031752 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10031752 Supplier Thermo Scientific™ Supplier No. CM0057B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cultivate molds and yeasts with typical morphology and pigmentation with this acidic medium.

Cultivate molds and yeasts with typical morphology and pigmentation with Thermo Scientific™ Oxoid™ Malt Extract Broth (Dehydrated). Mycological peptone in the medium gives a luxuriant growth of yeasts and molds with typical morphology and pigmentation. Due to the acidic nature of the medium, it inhibits the growth of bacteria resulting in improved isolation of yeasts and molds from materials heavily infected with bacteria.

Malt Extract Broth is a liquid medium recommended for the cultivation of molds and yeasts.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Malt Extract Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 25.0L medium
compliance-icons
Product Identifier
  • MALT EXTRACT BROTH
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304-IF INHALED:.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: Barley, malt

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.